1,3-Diacetylbenzene 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
1,3-Diacetylbenzene 99%
Catalog # ACRO170090050
Supplier: Thermo Fisher Scientific
CAS Number: 6781-42-6
Jump to:
Product Details & Documents
Synonyms: Benzene-1,3-bis(acetyl)
- CAS Number:6781-42-6
- Formula:C₁₀H₁₀O₂
- MDL Number:MFCD00008740
- Molecular Weight:162.19 g/mol
- Boiling Pt:150…155 °C (15 mmHg)
- Melting Pt:35…37 °C
- Flash Pt:>110 °C
- EINECS:229-842-9
Documents
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:28 to 33 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



