Order Entry
Export
ContactUsLinkComponent
4,6-Dichloropyrimidine 97%
Irritant
Corrosive
Environment
4,6-Dichloropyrimidine 97%
Certificates
undefined
Irritant
Corrosive
Environment
4,6-Dichloropyrimidine 97%

Specification Test Results

  • Appearance:
    White to yellow to orange Powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    64 to 70 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information