Order Entry
Export
ContactUsLinkComponent
Undec-10-enoic acid 99%
Irritant
Undec-10-enoic acid 99%
Certificates
undefined
Irritant
Undec-10-enoic acid 99%

Specification Test Results

  • Appearance:
    White to light yellow Low melting solid or liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.0 %
  • Titration with NaOH:
    98.5 to 101.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information