
4,4'-Difluorobenzophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:345-92-6
:
Formula:C₁₃H₈F₂O
MDL Number:MFCD00000353
Boiling Pt:200 °C (760 mmHg)
Melting Pt:106…109 °C
Density:1,39 g/cm³ (20 °C)
:218.2 g/mol
Storage Temperature:Ambient
EINECS:206-466-3
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:106 to 109 °C
- Loss on drying:≤0.25 %
- GC:≥98.5 %