Order Entry
Export
Orders LinkContactUsLinkComponent
Thiophene-2-carbaldehyde 98%

Thiophene-2-carbaldehyde 98%

Irritant
Thiophene-2-carbaldehyde 98%
Certificates
Thiophene-2-carbaldehyde 98%

3 Options Available Below

Product Details & Documents
Synonyms:  2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde
  • CAS Number:98-03-3
  • Formula:C₅H₄OS
  • MDL Number:MFCD00005429
  • Molecular Weight:112.15 g/mol
  • Boiling Pt:198 °C (760 mmHg)
  • Melting Pt:? 10 °C
  • Density:1.2 g/cm³
  • Flash Pt:77 °C
  • Storage Temperature:Refrigerator
  • EINECS:202-629-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear yellow to light brown Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5890 to 1.5920 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 32,20 €
    32,20 €
    1 * 25 mL
       32,20 €

  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 60,00 €
    60,00 €
    1 * 100 mL
       60,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 430,00 €
    430,00 €
    1 * 1 L
       430,00 €