
Oleyl alcohol ca. 60%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:143-28-2
:
Formula:C₁₈H₃₆O
MDL Number:MFCD00002993
:268.48 g/mol
Merck Index:12,06968
Boiling Pt:207 °C (13 mmHg)
Melting Pt:5 °C
Density:0.84 g/cm³
Flash Pt:170 °C
EINECS:205-597-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4590 to 1.4640 (20°C, 589 nm)
- GC:60 % Typical