Order Entry
Export
Orders LinkContactUsLinkComponent
2-Mercaptobenzothiazole 95%

2-Mercaptobenzothiazole 95%

Irritant
Environment
2-Mercaptobenzothiazole 95%
Certificates
2-Mercaptobenzothiazole 95%

4 Options Available Below

Product Details & Documents
Synonyms:  2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
  • CAS Number:149-30-4
  • Formula:C₇H₅NS₂
  • MDL Number:MFCD00005781
  • Molecular Weight:167.26 g/mol
  • Merck Index:13,05893
  • Boiling Pt:302 °C (1013 hPa)
  • Melting Pt:172…182 °C
  • Density:1,42 g/cm³ (20 °C)
  • Flash Pt:243 °C
  • Storage Temperature:Ambient
  • EINECS:205-736-8
  • UN:3077
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    172 to 180 °C
  • Loss on drying:
    ≤0.5 % (60 °C, 3 hrs)
  • Sulfated ash:
    ≤0.5 %
  • Titration with NaOH:
    ≥94 %
  • Impurity:
    Contains 1 to 2% oil
  • UV:
    at 250 nm A: ≤0.05
  • Non-volatile matter:
    ≤0.0003 %
  • Acidity:
    ≤0.0002 meq/g
  • Alkalinity:
    ≤0.0002 meq/g
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 22,40 €
    22,40 €
    1 * 10 g
       22,40 €

  • Plastic bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 21,40 €
    21,40 €
    1 * 50 g
       21,40 €

  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 28,20 €
    28,20 €
    1 * 250 g
       28,20 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 64,50 €
    64,50 €
    1 * 1 kg
       64,50 €