Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethyl propiolate 99%

Ethyl propiolate 99%
 
Ethyl propiolate 99%
Ethyl propiolate 99%

 

 
 :  Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
  •  :623-47-2
  •  :C₅H₆O₂
  •  :MFCD00009184
  •  :98.1 g/mol
  •  :120 °C (760 mmHg)
  •  :9 °C
  •  :0.968 g/cm³
  •  :23 °C
  •  :Refrigerator
  •  :210-795-8
  •  :1993
  •  :3,III

 

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4110 to 1.4130 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    5 g
       
       
    1 * 5 g
    1 * 5 g - 45,10 €
    45,10 €
    1 * 5 g
       45,10 €

  • Glass bottle for solids
    25 g
       
       
    1 * 25 g
    1 * 25 g - 155,00 €
    155,00 €
    1 * 25 g
       155,00 €

  • Glass bottle
    100 g
       
       
    1 * 100 g
    1 * 100 g - 442,00 €
    442,00 €
    1 * 100 g
       442,00 €