
4,4'-Dihydroxybenzophenone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:611-99-4
:
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00002358
:214.22 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:217…222 °C
Density:1,336 g/cm³ (20 °C)
EINECS:210-288-1
:GHS07
Specification Test Results
- Appearance:Off-white to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:217 to 222 °C
- GC:≥96.0 %
- Titration with NaOH:≥96.0 %