
2-Amino-4,6-dimethylpyrimidine 98%
Catalog # ACRO103590250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:150 to 154 °C
- Titration with HClO4:≥97.5 %
Specification Test Results
About this item
:
:123.16 g/mol
Melting Pt:150…154 °C
EINECS:212-181-5
:GHS07
CAS Number:767-15-7
Formula:C₆H₉N₃
MDL Number:MFCD00006102