
2-Amino-4,6-dichloropyrimidine 95% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-05-3
:
Formula:C₄H₃Cl₂N₃
MDL Number:MFCD00006090
:163.99 g/mol
Melting Pt:219…226 °C
EINECS:200-253-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange to light brown Powder
- Infrared spectrum:Conforms
- Melting point:216 to 226 °C
- HPLC:≥94 %