2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105)
Catalog # ACCUC-105N
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:326.44 g/mol
UN:3432
ADR:9,II
CAS Number:32598-14-4
Formula:C₁₂H₅Cl₅