2,3,4,5-Tetrachlorobiphenyl (PCB No. 61)
Catalog # ACCUC-061N
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:291.99 g/mol
UN:2315
ADR:9,II
CAS Number:33284-53-6
Formula:C₁₂H₆Cl₄