(R)-(-)-1-Phenyl-1,2-ethanediol, Sigma-Aldrich®
Catalog # SIAL302163-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:272 °C (1013 hPa)
Melting Pt:67…69 °C
:138.17 g/mol
CAS Number:16355-00-3
Formula:C₈H₁₀O₂
MDL Number:MFCD00064262