(R)-(-)-1-Phenyl-1,2-ethanediol ≥99%
Catalog # L10494.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:16355-00-3
:
Formula:C₈H₁₀O₂
Boiling Pt:272…274 °C
Melting Pt:67…69 °C
:138.17 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00064262