L(-)-Proline
Catalog # EHERC16345600
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:115.13 g/mol
Merck Index:13,07871
Boiling Pt:245 °C (1013 hPa)
Melting Pt:220…222 °C
Density:1.35 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:205-702-2
CAS Number:147-85-3
Formula:C₅H₉NO₂
MDL Number:MFCD00064318