Order Entry
Export
ContactUsLinkComponent
(R)-(-)-Mandelic acid
(R)-(-)-Mandelic acid
Catalog # MOLE17027682-25G
Supplier:  Molekula
CAS Number:  
(R)-(-)-Mandelic acid
Catalog # MOLE17027682-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :152.15 g/mol
Boiling Pt:322 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1,341 g/cm³ (20 °C)
Flash Pt:>190 °C
EINECS:210-276-6
 :GHS05
CAS Number:611-71-2
Formula:C₈H₈O₃
MDL Number:MFCD00064251