(R)-(-)-Mandelic acid 99%
:611-71-2
:
:C₈H₈O₃
:MFCD00064251
:152.15 g/mol
:322 °C (1013 hPa)
:130…135 °C
:1,341 g/cm³ (20 °C)
:>190 °C
:210-276-6
:GHS05
:Ambient
- Appearance:White to light yellow to light beige Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:130 to 135 °C
- Water:≤1 % (K.F.)
- Sulfated ash:≤0.2 %
- Specific optical rotation:-152° to -158° (20 °C, 589 nm) (c=2, H2O)
- Titration with NaOH:≥98.5 %