Order Entry
Export
ContactUsLinkComponent
L(+)-Glutamine, Sigma-Aldrich®
L(+)-Glutamine, Sigma-Aldrich®
Catalog # SIALM4655-500ML
Supplier:  Merck
CAS Number:  
undefined
L(+)-Glutamine, Sigma-Aldrich®
Catalog # SIALM4655-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :146.15 g/mol
Merck Index:13,04484
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044