Dipotassium-L(+)-tartrate hemihydrate, Sigma-Aldrich®
Catalog # SIAL25510-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:235.28 g/mol
Merck Index:12,07849
Density:1.943 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-697-7
CAS Number:6100-19-2
Formula:KOOC(CHOH)₂COOK·0.5H₂O
MDL Number:MFCD00150136