Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-3-fluorobenzene-1,2-diamine
5-Chloro-3-fluorobenzene-1,2-diamine 97%
5-Chloro-3-fluorobenzene-1,2-diamine 97%
:
Apollo Scientific
5-Chloro-3-fluorobenzene-1,2-diamine 97%
:
Apollo Scientific
:
1106717-48-9
:
:
C₆H₆ClFN₂
:
MFCD19441310
:
160.58 g/mol
:
GHS07