meso-2,3-Butanediol, Sigma-Aldrich®
Catalog # SIAL361461-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
Synonyms: meso-2,3-Dihydroxy butane, (R,S)-2,3-Butanediol, (R,S)-2,3-Butylene glykol, meso-2,3-Butylene glykol, Meso-2,3-butanediol, (R,S)-2,3-Dihydroxy butane
:
:90.12 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:5341-95-7
Formula:C₄H₁₀O₂
MDL Number:MFCD00077669