2,6-Dimethyl-4-nitroaniline 95%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:16947-63-0
- Formula:C₈H₁₀N₂O₂
- MDL Number:MFCD00211068
- Molecular Weight:166.18 g/mol
- Melting Pt:165 °C
3 Options Available Below
Product Details & Documents
- CAS Number:16947-63-0
- Formula:C₈H₁₀N₂O₂
- MDL Number:MFCD00211068
- Molecular Weight:166.18 g/mol
- Melting Pt:165 °C



