
5,8-Dihydroxy-1,4-naphthoquinone tech
Catalog # APOSOR01425-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.16 g/mol
:GHS07
CAS Number:475-38-7
Formula:C₁₀H₆O₄
MDL Number:MFCD00001685