Order Entry
Export
ContactUsLinkComponent
3-(2-Ethoxyphenoxy)-1,2-propanediol 95%
Irritant
3-(2-Ethoxyphenoxy)-1,2-propanediol 95%
Catalog # APOSOR62179-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(2-Ethoxyphenoxy)-1,2-propanediol 95%
Catalog # APOSOR62179-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.24 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:63834-83-3
Formula:C₁₁H₁₆O₄
MDL Number:MFCD00867665