Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethyl-4-nitroaniline
3,5-Dimethyl-4-nitroaniline 95%
3,5-Dimethyl-4-nitroaniline 95%
Irritant
3,5-Dimethyl-4-nitroaniline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dimethyl-4-nitroaniline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
34761-82-5
:
Formula:
C₈H₁₀N₂O₂
MDL Number:
MFCD00156608
:
166.18 g/mol
:
GHS07
Storage Temperature:
Refrigerator