Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Bis(phenylthio)thiophene
2,5-Bis(phenylthio)thiophene
2,5-Bis(phenylthio)thiophene
Irritant
2,5-Bis(phenylthio)thiophene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,5-Bis(phenylthio)thiophene
Supplier:
Apollo Scientific
About this item
CAS Number:
2974-09-6
:
Formula:
C₁₆H₁₂S₃
MDL Number:
MFCD02065045
:
300.46 g/mol
Melting Pt:
42…43 °C
:
GHS07
Storage Temperature:
Refrigerator