Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-1,3-Cyclohexanediol
trans-1,3-Cyclohexanediol 98%
trans-1,3-Cyclohexanediol 98%
APOSOR938906-1G
:
Apollo Scientific
:
trans-1,3-Cyclohexanediol 98%
APOSOR938906-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
116.16 g/mol
:
113 - 120 °C
:
GHS07
:
5515-64-0
:
C₆H₁₂O₂
:
MFCD16295186
:
113…120 °C