Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dichloropropane 99%

1,3-Dichloropropane 99%

Flammable
Supplier:   Apollo Scientific
1,3-Dichloropropane 99%
Supplier:  Apollo Scientific
Certificates
1,3-Dichloropropane 99%

2 Options Available Below

Product Details & Documents
Synonyms:  Trimethylendichloride
  • CAS Number:142-28-9
  • Formula:ClH₂CCH₂CH₂Cl
  • MDL Number:MFCD00000999
  • Molecular Weight:112.99 g/mol
  • Boiling Pt:120 °C
  • Density:1.19
  • Flash Pt:21 °C
  • UN:1933
  • ADR:3,II
  •  :Density: 1.1717 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 44,90 €
    44,90 €
    1 * 25 g
       44,90 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 89,80 €
    89,80 €
    1 * 100 g
       89,80 €