3,5-Dimethoxyphenol
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:500-99-2
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008388
- Molecular Weight:154.17 g/mol
- Boiling Pt:172…175/17 mm °C
- Boiling Pt:172 - 175/17 mm °C
- Melting Pt:40…45 °C
- Melting Pt:40 - 45 °C
- Flash Pt:78 °C
2 Options Available Below
Product Details & Documents
- CAS Number:500-99-2
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008388
- Molecular Weight:154.17 g/mol
- Boiling Pt:172…175/17 mm °C
- Boiling Pt:172 - 175/17 mm °C
- Melting Pt:40…45 °C
- Melting Pt:40 - 45 °C
- Flash Pt:78 °C



