Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3,6-difluorophenacyl bromide
2-Bromo-3,6-difluorophenacyl bromide 95%
2-Bromo-3,6-difluorophenacyl bromide 95%
Corrosive
2-Bromo-3,6-difluorophenacyl bromide 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
2-Bromo-3,6-difluorophenacyl bromide 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1807197-48-3
:
Formula:
C₈H₄Br₂F₂O
MDL Number:
MFCD28061331
:
313.92 g/mol
UN:
3265
ADR:
8,II
:
GHS05