
2-Nitro-m-xylene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:81-20-9
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007181
:151.16 g/mol
Boiling Pt:225…227 °C
Melting Pt:14…16 °C
Density:1.112
Flash Pt:88 °C
UN:1665
ADR:6.1,II
:GHS07