Ethyl-1,5-Dimethylpyrazole-3-carboxylate
Catalog # APOSOR29763-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:168.20 g/mol
Boiling Pt:154/10 mm °C
Melting Pt:42 - 46 °C
:GHS07
CAS Number:5744-51-4
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD00085071
Melting Pt:42…46 °C