

3-Oxa-9-azaspiro[5.5]undecane
Catalog # APOSOR917623-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:155.24 g/mol
:GHS05
:GHS07
CAS Number:311-21-7
Formula:C₉H₁₇NO
MDL Number:MFCD11505644