Order Entry
Export
ContactUsLinkComponent
Pesticide reference standard, 2,3,4-Trichloroanisole 10 µg/ml in isooctane, Dr Ehrenstorfer
Pesticide reference standard, 2,3,4-Trichloroanisole 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL17713400IO
 :  
Pesticide reference standard, 2,3,4-Trichloroanisole 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL17713400IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    54135-80-7
  • Description:
    2,3,4-Trichloroanisole 10 µg/ml in isooctane
  • Formula:
    C₇H₅Cl₃O
  • Description:
    2,3,4-Trichloroanisole