Pesticide reference standard, 1-Chloro-4-nitrobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11453700ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:100-00-5
- Description:1-Chloro-4-nitrobenzene 100 µg/ml in methanol
- Formula:C₆H₄ClNO₂
- Description:1-Chloro-4-nitrobenzene