(±)-Mandelic acid ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 90-64-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Physical Form: Cryst powder Sensitivity: Light Sensitive Appearance: White Melting Point: 118-121° Beilstein Reference: 510011 Merck Reference: 14,5717 MDL No.: MFCD00064250
Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
- CAS Number:90-64-2
- Formula:C₈H₈O₃
- MDL Number:MFCD00064250
- Molecular Weight:152.15 g/mol
- Merck Index:14,5717
- Boiling Pt:322 °C (1013 hPa)
- Melting Pt:118…121 °C
- Density:1.361 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-007-6
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 90-64-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Physical Form: Cryst powder Sensitivity: Light Sensitive Appearance: White Melting Point: 118-121° Beilstein Reference: 510011 Merck Reference: 14,5717 MDL No.: MFCD00064250
Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
- CAS Number:90-64-2
- Formula:C₈H₈O₃
- MDL Number:MFCD00064250
- Molecular Weight:152.15 g/mol
- Merck Index:14,5717
- Boiling Pt:322 °C (1013 hPa)
- Melting Pt:118…121 °C
- Density:1.361 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-007-6



