
(±)-4-Hydroxy-3-methoxymandelic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:55-10-7
:2394-20-9
Formula:C₉H₁₀O₅
MDL Number:MFCD00004235
:198.18 g/mol
Merck Index:12,10070
Boiling Pt:421 °C (1013 hPa)
Melting Pt:132…134 °C
Density:1.44 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-224-0
:GHS07
Storage Temperature:Refrigerator