Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene glycol 8,000
Polyethylene glycol 8,000, Sigma-Aldrich®
Polyethylene glycol 8,000, Sigma-Aldrich®
:
Merck
Polyethylene glycol 8,000, Sigma-Aldrich®
:
Merck
:
25322-68-3
:
MFCD00081839
:
H(OCH₂CH₂)
n
OH
:
MFCD01779615
:
8000 g/mol
:
12,07729
:
250 °C (1013 hPa)
:
1.125 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ200
:
Ambient
:
500-038-2