Polyethylene glycol 600, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
:600 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:17…22 °C
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:270 °C
Storage Temperature:Ambient
EINECS:500-038-2