Polyethylene glycol 2,000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
:2000 g/mol
Melting Pt:49…52 °C
Density:1.21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:500-038-2