Order Entry
Export
Orders LinkContactUsLinkComponent
 

DBCO-Cy3 fluorescent probe

Plastic bottle, 1 mg
Catalog # AATB920
Irritant
Supplier:   AAT Bioquest
DBCO-Cy3 fluorescent probe
Catalog # AATB920
Supplier:  AAT Bioquest
CAS Number:  1782950-79-1
DBCO-Cy3 fluorescent probe
DBCO-Cy3 fluorescent probe

Plastic bottle, 1 mg

CAS Number:  1782950-79-1
Price
156,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

This azadibenzocyclooctyne-cyanine dye derivative is a versatile labeling reagent for detection of azide containing molecules or compounds.

  • Copper-free reaction
  • Azide reactive
Synonyms:  2-[3-[1-[6-[[3-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]amino]-6-oxohexyl]-1,3-dihydro-3,3-dimethyl-5-sulpho-2H-indol-2-ylidene]-1-propen-1-yl]-3,3-dimethyl-5-sulpho-1-(3-sulphopropyl)-3H-indolium inner salt
  • CAS Number:1782950-79-1
  • MDL Number:MFCD28505574
  • Molecular Weight:1185.56 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications