Order Entry
Export
ContactUsLinkComponent
 
Irritant
Rhodamine aldehyde [5-TAMRA aldehyde] for biomolecular labeling
Catalog # AATB9005
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Rhodamine aldehyde [5-TAMRA aldehyde] for biomolecular labeling
Catalog # AATB9005
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :536.03 g/mol
ADR:5,II
 :GHS07