Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
CDPI3 Maleimide
CDPI3 Maleimide DNA binding
CDPI3 Maleimide DNA binding
CDPI3 Maleimide DNA binding
Catalog # AATB6908
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
CDPI3 Maleimide DNA binding
Catalog # AATB6908
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details