Rhodamine B ≥95%
Plastic bottle, 100 mg
Catalog # AATB69
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Rhodamine B (also called Rhodamine 610, Basic Violet 10, or C.I.
Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
- CAS Number:81-88-9
- Formula:C₂₈H₃₁ClN₂O₃
- MDL Number:MFCD00011931
- Molecular Weight:479.02 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mg



