Monochlorobimane (MCB) ≥95%
Plastic bottle, 10 mg
Catalog # AATB635
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Monochloromobimane (mBCl) is essentially nonfluorescent until reacted with a thiol group.
Synonyms: Chlorobimane, mBCI, 1H,7H-Pyrazolo(1,2-α)pyrazole-1,7-dione, 3-(chloromethyl)-2,5,6-trimethyl-
- CAS Number:76421-73-3
- Formula:C₁₀H₁₁ClN₂O₂
- MDL Number:MFCD00077379
- Molecular Weight:226.66 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:10 mg



