Bromobimane (mBBr) ≥95%
Plastic bottle, 25 mg
Catalog # AATB633
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
mBBr is one of the most popular thiol-reactive fluorescent tags, and is widely used to detect various thiol-containing biomolecules such as glutathione in cells.
Synonyms: Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione
- CAS Number:71418-44-5
- Formula:C₁₀H₁₁BrN₂O₂
- MDL Number:MFCD00036951
- Molecular Weight:271.11 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25 mg



