Pumice stone
Glass bottle for solids, 2,5 kg
Catalog # ROTH5832.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:1332-09-8
- MDL Number:MFCD00148147
- Melting Pt:> 500 °C
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 kg



