
Nile blue chloride (Basic blue 12) single isomer DNA stain
Catalog # AATB59
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:353.85 g/mol
ADR:5,II
:GHS07
Storage Temperature:Freezer
CAS Number:2381-85-3
Formula:C₂₀H₂₀ClN₃O
MDL Number:MFCD00011936