Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Texas Red-X acid
Texas Red-X acid fluorescent probe
Texas Red-X acid fluorescent probe
Texas Red-X acid fluorescent probe
AATB469
:
AAT BIOQUEST
:
Texas Red-X acid fluorescent probe
AATB469
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
10 mg
Seq:
2275
:
:
719.87 g/mol
:
5,II
:
GHS07
:
199745-67-0